Product Details

SNP ID
rs11101429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:48692049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAAGGGTGATGAAGCCCAGGAT[C/G]GGGGGTGTGAGCCTCAGGGCACAGG
Phenotype
MIM: 613316
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WDFY4 PubMed Links

Gene Details

Gene
WDFY4
Gene Name
WDFY family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020945.1 Intron NP_065996.1
XM_005270004.3 Intron XP_005270061.1
XM_011539986.2 Intron XP_011538288.2
XM_011539987.2 Intron XP_011538289.2
XM_011539988.2 Intron XP_011538290.1
XM_011539990.2 Intron XP_011538292.1
XM_011539991.2 Intron XP_011538293.1
XM_011539992.2 Intron XP_011538294.1
XM_017016463.1 Intron XP_016871952.1
XM_017016464.1 Intron XP_016871953.1
XM_017016465.1 Intron XP_016871954.1

View Full Product Details