Product Details
- SNP ID
-
rs2527901
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:99927489 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGTGTAGCAGGAAGAAAAAAAAA[A/T]TTTCCTAGCTATTGCCAAAGCATCC
- Phenotype
-
MIM: 611925
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GJC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78828444] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GJC3
- Gene Name
- gap junction protein gamma 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181538.2 |
|
Intron |
|
|
NP_853516.1 |
- Gene
- LOC101927610
- Gene Name
- uncharacterized LOC101927610
There are no transcripts associated with this gene.
- Gene
- TRIM4
- Gene Name
- tripartite motif containing 4
There are no transcripts associated with this gene.
View Full Product Details