Product Details

SNP ID
rs2728
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:154962607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTTCTAGTCCTAGATGGAAAT[A/C]AATTTCATCTGCCTCAGGTCTCTGG
Phenotype
MIM: 606903 MIM: 600560
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928120 PubMed Links

Gene Details

Gene
LOC101928120
Gene Name
uncharacterized LOC101928120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321726.1 1137 UTR 3 NP_001308655.1
Gene
PBXIP1
Gene Name
PBX homeobox interacting protein 1
There are no transcripts associated with this gene.

Gene
PYGO2
Gene Name
pygopus family PHD finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138300.3 1137 Intron NP_612157.1
Gene
SHC1
Gene Name
SHC adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130040.1 1137 Intron NP_001123512.1
NM_001130041.1 1137 Intron NP_001123513.1
NM_001202859.1 1137 Intron NP_001189788.1
NM_003029.4 1137 Intron NP_003020.2
NM_183001.4 1137 Intron NP_892113.4
XM_005245449.4 1137 Intron XP_005245506.1
XM_005245451.4 1137 Intron XP_005245508.1
XM_011509892.2 1137 UTR 3 XP_011508194.1
XM_011509893.2 1137 UTR 3 XP_011508195.1
XM_011509894.2 1137 UTR 3 XP_011508196.1
XM_011509897.1 1137 Intron XP_011508199.1
XM_011509898.2 1137 Intron XP_011508200.1
XM_017002081.1 1137 UTR 3 XP_016857570.1
XM_017002082.1 1137 UTR 3 XP_016857571.1
XM_017002083.1 1137 Intron XP_016857572.1

View Full Product Details