Product Details

SNP ID
rs2448113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26698377 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGACCTTAGTTGTCCAGCAGTGGC[C/T]TCGGGAGGAGAGCACACCTGGATGG
Phenotype
MIM: 607429
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDSS1 PubMed Links

Gene Details

Gene
PDSS1
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321978.1 Intron NP_001308907.1
NM_001321979.1 Intron NP_001308908.1
NM_014317.4 Intron NP_055132.2
XM_011519437.2 Intron XP_011517739.1
XM_017016011.1 Intron XP_016871500.1

View Full Product Details