Product Details
- SNP ID
-
rs12776891
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:26704425 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGATAGATATGAATATGGAAATG[A/G]CTTTCTTTGGGGGAAGGGATGGACT
- Phenotype
-
MIM: 607429
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PDSS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143530252] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PDSS1
- Gene Name
- prenyl (decaprenyl) diphosphate synthase, subunit 1
View Full Product Details