Product Details

SNP ID
rs10852767
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75991571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCAAGTTGGTGGAGCCCTTCCA[C/T]GCCCAGGTGGGCTCCCTGTACATCG
Phenotype
MIM: 123828 MIM: 613130
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CDK3 PubMed Links

Gene Details

Gene
CDK3
Gene Name
cyclin dependent kinase 3
There are no transcripts associated with this gene.

Gene
TEN1
Gene Name
TEN1 CST complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113324.2 499 Silent Mutation CAC,CAT H66H NP_001106795.2
Gene
TEN1-CDK3
Gene Name
TEN1-CDK3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details