Product Details
- SNP ID
-
hCV26215844
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:51332445 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGTGAATTTCGCTGTGCGCCAAG[A/G]AGGTTCCTGGCCCCAGGAGTGCGAA
- Phenotype
-
MIM: 300527
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUDT10
PubMed Links
Gene Details
- Gene
- NUDT10
- Gene Name
- nudix hydrolase 10
There are no transcripts associated with this gene.
View Full Product Details