Product Details

SNP ID
rs2940531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176906768 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGAATGATGCACAAAACAATAG[A/G]AGGAAGCAATAATGTCTGCACATCT
Phenotype
MIM: 142570 MIM: 609433
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HK3 PubMed Links

Gene Details

Gene
HK3
Gene Name
hexokinase 3
There are no transcripts associated with this gene.

Gene
UIMC1
Gene Name
ubiquitin interaction motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199297.2 Intron NP_001186226.1
NM_001199298.1 Intron NP_001186227.1
NM_001317961.1 Intron NP_001304890.1
NM_016290.4 Intron NP_057374.3
XM_005265930.2 Intron XP_005265987.1
XM_005265932.2 Intron XP_005265989.1
XM_005265933.2 Intron XP_005265990.1
XM_005265934.2 Intron XP_005265991.1
XM_005265935.2 Intron XP_005265992.1
XM_005265936.2 Intron XP_005265993.1
XM_006714871.2 Intron XP_006714934.1
XM_006714874.2 Intron XP_006714937.1
XM_011534568.2 Intron XP_011532870.1
XM_011534570.1 Intron XP_011532872.1
XM_017009574.1 Intron XP_016865063.1
XM_017009575.1 Intron XP_016865064.1
XM_017009576.1 Intron XP_016865065.1
XM_017009577.1 Intron XP_016865066.1
XM_017009578.1 Intron XP_016865067.1
XM_017009579.1 Intron XP_016865068.1

View Full Product Details