Product Details

SNP ID
rs36040764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50548657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGGCGCCGGGGGAGGGGGGCA[A/G]CCCTGCCGGCCGCAGCGGGGCGCTC
Phenotype
MIM: 615775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHDC7B PubMed Links

Gene Details

Gene
KLHDC7B
Gene Name
kelch domain containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138433.3 625 Missense Mutation AAC,AGC N164S NP_612442.2
Gene
SYCE3
Gene Name
synaptonemal complex central element protein 3
There are no transcripts associated with this gene.

View Full Product Details