Product Details

SNP ID
rs4660375
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:40045467 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTGGAGTACAATGGTGCAATCA[C/T]GGCTGATTGCAACCTCCAACTTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAP1 PubMed Links

Gene Details

Gene
CAP1
Gene Name
adenylate cyclase associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105530.1 Intron NP_001099000.1
NM_006367.3 Intron NP_006358.1
XM_005270368.1 Intron XP_005270425.1
XM_011540509.1 Intron XP_011538811.1
XM_011540510.1 Intron XP_011538812.1
XM_011540511.1 Intron XP_011538813.1
XM_011540512.1 Intron XP_011538814.1
XM_011540513.2 Intron XP_011538815.1
XM_011540514.1 Intron XP_011538816.1
XM_011540515.1 Intron XP_011538817.1
XM_017000067.1 Intron XP_016855556.1
XM_017000068.1 Intron XP_016855557.1
XM_017000069.1 Intron XP_016855558.1
XM_017000070.1 Intron XP_016855559.1
XM_017000071.1 Intron XP_016855560.1
XM_017000072.1 Intron XP_016855561.1
XM_017000073.1 Intron XP_016855562.1

View Full Product Details