Product Details

SNP ID
rs1591294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54621372 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTTCTTAAAAGTAGCCATTTTGA[C/T]TGGTGTAAGTTGATACTCAGTGTGG
Phenotype
MIM: 604811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LILRB1 PubMed Links

Gene Details

Gene
LILRB1
Gene Name
leukocyte immunoglobulin like receptor B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081637.2 Intron NP_001075106.2
NM_001081638.3 Intron NP_001075107.2
NM_001081639.3 Intron NP_001075108.2
NM_001278398.2 Intron NP_001265327.2
NM_001278399.2 Intron NP_001265328.2
NM_006669.6 Intron NP_006660.4
XM_011526331.2 Intron XP_011524633.1
XM_011526332.2 Intron XP_011524634.1
XM_011526335.2 Intron XP_011524637.1
XM_011526336.2 Intron XP_011524638.1
XM_011526338.2 Intron XP_011524640.1
XM_017026182.1 Intron XP_016881671.1
XM_017026183.1 Intron XP_016881672.1
XM_017026184.1 Intron XP_016881673.1
XM_017026185.1 Intron XP_016881674.1
XM_017026186.1 Intron XP_016881675.1
XM_017026187.1 Intron XP_016881676.1
XM_017026188.1 Intron XP_016881677.1
XM_017026189.1 Intron XP_016881678.1
XM_017026190.1 Intron XP_016881679.1
XM_017026191.1 Intron XP_016881680.1
XM_017026192.1 Intron XP_016881681.1

View Full Product Details