Product Details

SNP ID
rs34937940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55580308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGAAATAGAGTGTGTTCTCAGA[C/T]GCCAGGGGCTGGGAGAGGTCGGGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF579 PubMed Links

Gene Details

Gene
ZNF579
Gene Name
zinc finger protein 579
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152600.2 Intron NP_689813.2
XM_017026409.1 Intron XP_016881898.1
XM_017026410.1 Intron XP_016881899.1
XM_017026411.1 Intron XP_016881900.1

View Full Product Details