Product Details

SNP ID
rs11643147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89972239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCAGAGAACAACCGACCAAGC[A/G]GACAAACGGAACTCCGTGAGGGGGA
Phenotype
MIM: 603020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFG3L1P PubMed Links

Gene Details

Gene
AFG3L1P
Gene Name
AFG3 like matrix AAA peptidase subunit 1, pseudogene
There are no transcripts associated with this gene.

Gene
CENPBD1
Gene Name
CENPB DNA-binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145039.3 594 UTR 5 NP_659476.2
Gene
DEF8
Gene Name
differentially expressed in FDCP 8 homolog (mouse)
There are no transcripts associated with this gene.

View Full Product Details