Product Details

SNP ID
rs5023650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38563658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCATGTCTGTTAGCCTTTCAGA[A/C]ATATAAAATCAGTTAAATCTGTAGT
Phenotype
MIM: 611208
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPLL PubMed Links

Gene Details

Gene
HNRNPLL
Gene Name
heterogeneous nuclear ribonucleoprotein L like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142650.1 2654 UTR 3 NP_001136122.1
NM_138394.3 2654 UTR 3 NP_612403.2
XM_005264639.3 2654 UTR 3 XP_005264696.2
XM_005264640.3 2654 Intron XP_005264697.2
XM_011533165.2 2654 Intron XP_011531467.1
XM_011533166.2 2654 Intron XP_011531468.1

View Full Product Details