Product Details
- SNP ID
-
rs11642030
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:69571308 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACAGAGTGAGACTCTGTCTCAAAA[A/C]AACAACAACAAACAAACAAACAAAC
- Phenotype
-
MIM: 604708
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR1538
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141981673] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR1538
- Gene Name
- microRNA 1538
There are no transcripts associated with this gene.
- Gene
- NFAT5
- Gene Name
- nuclear factor of activated T-cells 5, tonicity-responsive
View Full Product Details