Product Details

SNP ID
rs34585481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:132252862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGGGAGGAACCCAGACCGTGA[A/T]GGTGGCCACAGGAACTCGGCTGCAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD30BL PubMed Links

Gene Details

Gene
ANKRD30BL
Gene Name
ankyrin repeat domain 30B-like
There are no transcripts associated with this gene.

Gene
MIR663B
Gene Name
microRNA 663b
There are no transcripts associated with this gene.

View Full Product Details