Product Details

SNP ID
rs10917571
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:161549621 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCATCCCTTCGTGGGAGTCTCATT[A/C]GTAGCCTGAAAAGGGGTCTCTGCTG
Phenotype
MIM: 146740
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FCGR3A PubMed Links

Gene Details

Gene
FCGR3A
Gene Name
Fc fragment of IgG receptor IIIa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127593.1 Intron NP_001121065.1
NM_001127595.1 Intron NP_001121067.1
NM_001127596.1 Intron NP_001121068.1
XM_011509293.2 Intron XP_011507595.2

View Full Product Details