Product Details

SNP ID
rs442914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49953470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGCTAAGTCGATTAGGAAAAGG[A/G]AACTGGAGGTGGCGAGAGTGGCTTA
Phenotype
MIM: 607157
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC11 PubMed Links

Gene Details

Gene
SIGLEC11
Gene Name
sialic acid binding Ig like lectin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135163.1 Intron NP_001128635.1
NM_052884.2 Intron NP_443116.2
XM_005258476.3 Intron XP_005258533.2
XM_005258477.3 Intron XP_005258534.2

View Full Product Details