Product Details

SNP ID
rs238004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:35480177 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTACCCCTTTTCTTACCCTTCCT[C/T]GGTCATCCTGGGAGCTCCACTTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRG2DP PubMed Links

Gene Details

Gene
FRG2DP
Gene Name
FSHD region gene 2 family member D, pseudogene
There are no transcripts associated with this gene.

View Full Product Details