Product Details

SNP ID
rs1684102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:35506579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGGGCGCGCCCAAAGTCTGGCG[A/G]AAGAGCGTTACCCTGGCAACCCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371204 PubMed Links

Gene Details

Gene
LOC105371204
Gene Name
uncharacterized LOC105371204
There are no transcripts associated with this gene.

Gene
TP53TG3HP
Gene Name
TP53 target 3 family member H, pseudogene
There are no transcripts associated with this gene.

View Full Product Details