Product Details

SNP ID
rs6945922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6015749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACAAAAACAAAACTAGCTTTCT[A/G]GTTTTCAAAGGCAGAGCCAAGAGCA
Phenotype
MIM: 600859 MIM: 613635 MIM: 600259
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIMP2 PubMed Links

Gene Details

Gene
AIMP2
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326606.1 Intron NP_001313535.1
NM_001326607.1 Intron NP_001313536.1
NM_001326609.1 Intron NP_001313538.1
NM_001326611.1 Intron NP_001313540.1
NM_006303.3 Intron NP_006294.2
XM_005249847.3 Intron XP_005249904.1
XM_005249848.2 Intron XP_005249905.1
XM_005249850.3 Intron XP_005249907.1
XM_006715772.2 Intron XP_006715835.1
XM_011515519.1 Intron XP_011513821.1
Gene
EIF2AK1
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 1
There are no transcripts associated with this gene.

Gene
PMS2
Gene Name
PMS1 homolog 2, mismatch repair system component
There are no transcripts associated with this gene.

View Full Product Details