Product Details

SNP ID
rs2951315
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144110425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCCTGTTTCACAGTGTCTTTAA[C/T]CCAATGCTGAACCCCCTGATCTACA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR2A2 PubMed Links

Gene Details

Gene
OR2A2
Gene Name
olfactory receptor family 2 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005480.2 843 Silent Mutation AAC,AAT N281N NP_001005480.2

View Full Product Details