Product Details

SNP ID
rs13165869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43490365 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTCAGGATGAATTTGTCACATA[A/G]AAAGAGCCACTCATTTTCATGTTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5orf34 PubMed Links

Gene Details

Gene
C5orf34
Gene Name
chromosome 5 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198566.3 Intron NP_940968.1
XM_011514038.2 Intron XP_011512340.1
XM_017009445.1 Intron XP_016864934.1
XM_017009446.1 Intron XP_016864935.1
Gene
TMEM267
Gene Name
transmembrane protein 267
There are no transcripts associated with this gene.

View Full Product Details