Product Details

SNP ID
rs2912798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:121481809 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATCATTAAAATAGAACTTTAGT[A/G]CTTTTCCCGGTTTCTTTCTTTTTTA
Phenotype
MIM: 176943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGFR2 PubMed Links

Gene Details

Gene
FGFR2
Gene Name
fibroblast growth factor receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000141.4 Intron NP_000132.3
NM_001144913.1 Intron NP_001138385.1
NM_001144914.1 Intron NP_001138386.1
NM_001144915.1 Intron NP_001138387.1
NM_001144916.1 Intron NP_001138388.1
NM_001144917.1 Intron NP_001138389.1
NM_001144918.1 Intron NP_001138390.1
NM_001144919.1 Intron NP_001138391.1
NM_001320654.1 Intron NP_001307583.1
NM_001320658.1 Intron NP_001307587.1
NM_022970.3 Intron NP_075259.4
NM_023029.2 Intron NP_075418.1
XM_006717708.2 Intron XP_006717771.1
XM_006717710.3 Intron XP_006717773.1
XM_006717711.2 Intron XP_006717774.1
XM_006717712.2 Intron XP_006717775.1
XM_017015920.1 Intron XP_016871409.1
XM_017015921.1 Intron XP_016871410.1
XM_017015922.1 Intron XP_016871411.1
XM_017015923.1 Intron XP_016871412.1
XM_017015924.1 Intron XP_016871413.1
XM_017015925.1 Intron XP_016871414.1

View Full Product Details