Product Details

SNP ID
rs11158071
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19976331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGACTGCTCACATCACTGTGGT[C/T]ACTTTATTCTTTGGACCATGCATTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR4K15 PubMed Links

Gene Details

Gene
OR4K15
Gene Name
olfactory receptor family 4 subfamily K member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005486.1 813 Silent Mutation GTC,GTT V271V NP_001005486.1

View Full Product Details