Product Details

SNP ID
rs2873723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19781355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAATGCTGCATTCACTTCCTCC[G/T]TTCATTTGTGTTCTTAAAATTTTAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4M1 PubMed Links

Gene Details

Gene
OR4M1
Gene Name
olfactory receptor family 4 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005500.1 Intron NP_001005500.1
XM_017021338.1 Intron XP_016876827.1
XM_017021339.1 Intron XP_016876828.1

View Full Product Details