Product Details

SNP ID
rs2732
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.1:6223538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTGATAAAATGCTTTTCTGAAC[A/G]TACATTTTAGGTATCTGGCACAATT
Phenotype
MIM: 605851 MIM: 616923
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ICMT PubMed Links

Gene Details

Gene
ICMT
Gene Name
isoprenylcysteine carboxyl methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012405.3 2170 UTR 3 NP_036537.1
XM_011541140.2 2170 UTR 3 XP_011539442.1
Gene
RNF207
Gene Name
ring finger protein 207
There are no transcripts associated with this gene.

View Full Product Details