Product Details
- SNP ID
-
rs1044154
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:6264300 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCAAGCAGCAGCTTTATTAGTGG[T/G]GCTGGGTTCTTCCCATACCCTACAG
- Phenotype
-
MIM: 602587
MIM: 614269
- Polymorphism
- T/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ACOT7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77120239] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACOT7
- Gene Name
- acyl-CoA thioesterase 7
- Gene
- GPR153
- Gene Name
- G protein-coupled receptor 153
There are no transcripts associated with this gene.
View Full Product Details