Product Details

SNP ID
rs12741781
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:243264850 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGTTGGTGCTGGGGAAAAGAAA[G/T]CAGGATGCATTCATTGTGAAATACC
Phenotype
MIM: 613023 MIM: 613524
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP170 PubMed Links

Gene Details

Gene
CEP170
Gene Name
centrosomal protein 170
There are no transcripts associated with this gene.

Gene
SDCCAG8
Gene Name
serologically defined colon cancer antigen 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006642.3 Intron NP_006633.1
XM_005273013.4 Intron XP_005273070.1
XM_005273018.1 Intron XP_005273075.1
XM_005273021.4 Intron XP_005273078.1
XM_005273022.3 Intron XP_005273079.1
XM_005273023.4 Intron XP_005273080.1
XM_006711727.3 Intron XP_006711790.1
XM_006711728.3 Intron XP_006711791.1
XM_011544021.2 Intron XP_011542323.1
XM_011544022.2 Intron XP_011542324.1
XM_011544023.2 Intron XP_011542325.1
XM_011544024.2 Intron XP_011542326.1
XM_011544026.2 Intron XP_011542328.1
XM_011544028.2 Intron XP_011542330.1
XM_011544029.2 Intron XP_011542331.1
XM_011544030.2 Intron XP_011542332.1
XM_017000103.1 Intron XP_016855592.1
XM_017000104.1 Intron XP_016855593.1
XM_017000105.1 Intron XP_016855594.1
XM_017000106.1 Intron XP_016855595.1
XM_017000107.1 Intron XP_016855596.1
XM_017000108.1 Intron XP_016855597.1
XM_017000109.1 Intron XP_016855598.1

View Full Product Details