Product Details

SNP ID
rs6695762
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:245164071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTGTATATATTTTTTTCTATTAA[A/G]ATGTTGGCTTTTGGACTGAGATCTG
Phenotype
MIM: 614026
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF26B PubMed Links

Gene Details

Gene
KIF26B
Gene Name
kinesin family member 26B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018012.3 Intron NP_060482.2

View Full Product Details