Product Details
- SNP ID
-
rs3733023
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:172633216 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTGCTTTCTGTAGCTGTAGGTAT[C/G]AGTATAGTTGGCTAAGATAACAAGA
- Phenotype
-
MIM: 613234
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NCEH1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3733022] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NCEH1
- Gene Name
- neutral cholesterol ester hydrolase 1
View Full Product Details