Product Details

SNP ID
rs7641929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:156811605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACCTTTATTGTAGTAGCCCCAA[C/G]GTGTGGAAGTTGTTTCCTGTGACCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00886 PubMed Links

Gene Details

Gene
LINC00886
Gene Name
long intergenic non-protein coding RNA 886
There are no transcripts associated with this gene.

Gene
PA2G4P4
Gene Name
proliferation-associated 2G4 pseudogene 4
There are no transcripts associated with this gene.

View Full Product Details