Product Details

SNP ID
rs10105470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:47314761 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACCATATCAGGATGTAAAGAAAA[C/G]AAATAGCAAACTTACAAACCTAGGT
Phenotype
MIM: 615384
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPIDR PubMed Links

Gene Details

Gene
SPIDR
Gene Name
scaffolding protein involved in DNA repair
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080394.3 Intron NP_001073863.1
NM_001282916.1 Intron NP_001269845.1
NM_001282919.1 Intron NP_001269848.1
XM_005251189.3 Intron XP_005251246.1
XM_005251191.3 Intron XP_005251248.1
XM_005251193.3 Intron XP_005251250.1
XM_005251195.4 Intron XP_005251252.1
XM_005251198.4 Intron XP_005251255.1
XM_005251199.4 Intron XP_005251256.1
XM_006716443.3 Intron XP_006716506.1
XM_006716444.3 Intron XP_006716507.1
XM_011517497.2 Intron XP_011515799.1
XM_011517500.2 Intron XP_011515802.1
XM_011517507.2 Intron XP_011515809.1
XM_017013268.1 Intron XP_016868757.1
XM_017013269.1 Intron XP_016868758.1
XM_017013270.1 Intron XP_016868759.1
XM_017013271.1 Intron XP_016868760.1
XM_017013272.1 Intron XP_016868761.1
XM_017013273.1 Intron XP_016868762.1
XM_017013274.1 Intron XP_016868763.1
XM_017013275.1 Intron XP_016868764.1
XM_017013276.1 Intron XP_016868765.1
XM_017013277.1 Intron XP_016868766.1
XM_017013278.1 Intron XP_016868767.1
XM_017013279.1 Intron XP_016868768.1
XM_017013280.1 Intron XP_016868769.1
XM_017013281.1 Intron XP_016868770.1
XM_017013282.1 Intron XP_016868771.1
XM_017013283.1 Intron XP_016868772.1
XM_017013284.1 Intron XP_016868773.1
XM_017013285.1 Intron XP_016868774.1
XM_017013286.1 Intron XP_016868775.1
XM_017013287.1 Intron XP_016868776.1
XM_017013288.1 Intron XP_016868777.1
XM_017013289.1 Intron XP_016868778.1
XM_017013290.1 Intron XP_016868779.1
XM_017013291.1 Intron XP_016868780.1

View Full Product Details