Product Details

SNP ID
rs1008250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133532712 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGAGCCCCTGGCATCAGGAGCT[A/G]CCAGAGCACCTGCTACGTGCCAGTC
Phenotype
MIM: 612277 MIM: 615345
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTSL2 PubMed Links

Gene Details

Gene
ADAMTSL2
Gene Name
ADAMTS like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145320.1 Intron NP_001138792.1
NM_014694.3 Intron NP_055509.2
XM_005272237.3 Intron XP_005272294.1
XM_005272238.3 Intron XP_005272295.1
XM_005272239.2 Intron XP_005272296.1
XM_011519241.2 Intron XP_011517543.2
XM_011519242.2 Intron XP_011517544.1
Gene
TMEM8C
Gene Name
transmembrane protein 8C
There are no transcripts associated with this gene.

View Full Product Details