Product Details

SNP ID
rs11807466
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:201960112 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTATGGACTTCAGTTAATAGTAA[C/T]GTGTCGGCCGGGCACAGTGGCTCAC
Phenotype
MIM: 605057
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIMM17A PubMed Links

Gene Details

Gene
TIMM17A
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006335.2 Intron NP_006326.1

View Full Product Details