Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004586.2 | 4866 | UTR 3 | NP_004577.1 | ||
XM_005274573.2 | 4866 | UTR 3 | XP_005274630.1 | ||
XM_005274577.2 | 4866 | UTR 3 | XP_005274634.1 | ||
XM_006724507.3 | 4866 | UTR 3 | XP_006724570.1 | ||
XM_011545555.1 | 4866 | Intron | XP_011543857.1 | ||
XM_011545556.1 | 4866 | UTR 3 | XP_011543858.1 | ||
XM_011545557.2 | 4866 | UTR 3 | XP_011543859.1 | ||
XM_011545558.2 | 4866 | UTR 3 | XP_011543860.1 | ||
XM_011545559.1 | 4866 | UTR 3 | XP_011543861.1 | ||
XM_011545560.1 | 4866 | UTR 3 | XP_011543862.1 | ||
XM_011545561.1 | 4866 | UTR 3 | XP_011543863.1 | ||
XM_011545562.1 | 4866 | UTR 3 | XP_011543864.1 | ||
XM_011545563.2 | 4866 | UTR 3 | XP_011543865.1 | ||
XM_017029713.1 | 4866 | UTR 3 | XP_016885202.1 | ||
XM_017029714.1 | 4866 | UTR 3 | XP_016885203.1 | ||
XM_017029715.1 | 4866 | UTR 3 | XP_016885204.1 | ||
XM_017029716.1 | 4866 | UTR 3 | XP_016885205.1 | ||
XM_017029717.1 | 4866 | UTR 3 | XP_016885206.1 | ||
XM_017029718.1 | 4866 | UTR 3 | XP_016885207.1 | ||
XM_017029719.1 | 4866 | UTR 3 | XP_016885208.1 |