Product Details

SNP ID
rs1136247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:20154468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTTAAAGGGCTTTGACGTATAT[G/T]GGCAAAATCCCAGATGATATGCCAA
Phenotype
MIM: 300075
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RPS6KA3 PubMed Links

Gene Details

Gene
RPS6KA3
Gene Name
ribosomal protein S6 kinase A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004586.2 3336 UTR 3 NP_004577.1
XM_005274573.2 3336 UTR 3 XP_005274630.1
XM_005274577.2 3336 UTR 3 XP_005274634.1
XM_006724507.3 3336 UTR 3 XP_006724570.1
XM_011545555.1 3336 Intron XP_011543857.1
XM_011545556.1 3336 UTR 3 XP_011543858.1
XM_011545557.2 3336 UTR 3 XP_011543859.1
XM_011545558.2 3336 UTR 3 XP_011543860.1
XM_011545559.1 3336 UTR 3 XP_011543861.1
XM_011545560.1 3336 UTR 3 XP_011543862.1
XM_011545561.1 3336 UTR 3 XP_011543863.1
XM_011545562.1 3336 UTR 3 XP_011543864.1
XM_011545563.2 3336 UTR 3 XP_011543865.1
XM_017029713.1 3336 UTR 3 XP_016885202.1
XM_017029714.1 3336 UTR 3 XP_016885203.1
XM_017029715.1 3336 UTR 3 XP_016885204.1
XM_017029716.1 3336 UTR 3 XP_016885205.1
XM_017029717.1 3336 UTR 3 XP_016885206.1
XM_017029718.1 3336 UTR 3 XP_016885207.1
XM_017029719.1 3336 UTR 3 XP_016885208.1

View Full Product Details