Product Details
- SNP ID
-
rs35902415
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:27311414 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGTACATTTTGGCCCTGTTCCCT[C/G]TGAAGCGTTCCATATTTTCCCCTGT
- Phenotype
-
MIM: 137960
MIM: 606474
MIM: 600945
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MPV17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147244564,rs199729168] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MPV17
- Gene Name
- MPV17, mitochondrial inner membrane protein
- Gene
- TRIM54
- Gene Name
- tripartite motif containing 54
There are no transcripts associated with this gene.
- Gene
- UCN
- Gene Name
- urocortin
There are no transcripts associated with this gene.
View Full Product Details