Product Details

SNP ID
rs2229048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111046994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGTCCTTCAGGTTTGGGAAGGC[A/G]TCCAACCACTTGGGCTCAAATACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTM2P1 PubMed Links

Gene Details

Gene
GSTM2P1
Gene Name
glutathione S-transferase mu 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details