Product Details

SNP ID
rs2289081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20682080 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGTCCACAGGAAGCTCCCGTGC[C/G]TGGCATAAGACCAGGTGGCTATGGA
Phenotype
MIM: 613570
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LDAH PubMed Links

Gene Details

Gene
LDAH
Gene Name
lipid droplet associated hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282719.1 Intron NP_001269648.1
NM_001282720.1 Intron NP_001269649.1
NM_001282721.1 Intron NP_001269650.1
NM_001282722.1 Intron NP_001269651.1
NM_001282723.1 Intron NP_001269652.1
NM_001282724.1 Intron NP_001269653.1
NM_021925.3 Intron NP_068744.1
XM_011533014.2 Intron XP_011531316.1

View Full Product Details