Product Details

SNP ID
rs11066180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112136906 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCATCTGGCCAAAACTGGCCAAC[A/G]TTGAATGAGTATGTTAATTCACCAA
Phenotype
MIM: 613197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAFD1 PubMed Links

Gene Details

Gene
TRAFD1
Gene Name
TRAF-type zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143906.1 Intron NP_001137378.1
NM_006700.2 Intron NP_006691.1

View Full Product Details