Product Details

SNP ID
rs2257984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14147647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTTCCAGAACCTGGACACAGGC[A/G]ATTCAAGACAGAGGCAGGTGTGGGC
Phenotype
MIM: 612048 MIM: 613208
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM43 PubMed Links

Gene Details

Gene
TMEM43
Gene Name
transmembrane protein 43
There are no transcripts associated with this gene.

Gene
XPC
Gene Name
XPC complex subunit, DNA damage recognition and repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004628.4 Intron NP_004619.3
XM_011534093.2 Intron XP_011532395.1

View Full Product Details