Product Details

SNP ID
rs2290314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12836078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTACATTTATTTAAACAGAAAA[C/T]GTGCACATGAGCTGCCTACTCATTT
Phenotype
MIM: 610403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAND2 PubMed Links

Gene Details

Gene
CAND2
Gene Name
cullin associated and neddylation dissociated 2 (putative)
There are no transcripts associated with this gene.

Gene
RPL32
Gene Name
ribosomal protein L32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000994.3 637 UTR 3 NP_000985.1
NM_001007073.1 637 UTR 3 NP_001007074.1
NM_001007074.1 637 UTR 3 NP_001007075.1
Gene
SNORA7A
Gene Name
small nucleolar RNA, H/ACA box 7A
There are no transcripts associated with this gene.

View Full Product Details