Product Details

SNP ID
rs7669660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99071451 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCAACTGGAGAACAGATAAATTA[C/T]GGTAGTTCCCGTAATTGAATTCTGC
Phenotype
MIM: 103710 MIM: 610151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADH5 PubMed Links

Gene Details

Gene
ADH5
Gene Name
alcohol dehydrogenase 5 (class III), chi polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000671.4 Intron NP_000662.3
Gene
METAP1
Gene Name
methionyl aminopeptidase 1
There are no transcripts associated with this gene.

View Full Product Details