Product Details

SNP ID
rs6656934
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:39501046 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGAGCCTCAGGCTACTTCAATTC[A/C]TGGCAGAAAGTGGAAGGACAGTGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BMP8A PubMed Links

Gene Details

Gene
BMP8A
Gene Name
bone morphogenetic protein 8a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181809.3 Intron NP_861525.2
XM_006710616.3 Intron XP_006710679.1
XM_011541381.2 Intron XP_011539683.1
XM_011541382.2 Intron XP_011539684.1
XM_017001198.1 Intron XP_016856687.1
Gene
LOC105378950
Gene Name
mitogen-activated protein kinase 7-like
There are no transcripts associated with this gene.

View Full Product Details