Product Details

SNP ID
rs2292836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96227385 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCCATAGTGCTCGTGTGATGTAC[A/C]CTAAGTTGTAAAGTTATAGGGCATC
Phenotype
MIM: 191330
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UQCRB PubMed Links

Gene Details

Gene
UQCRB
Gene Name
ubiquinol-cytochrome c reductase binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199975.2 4224 UTR 3 NP_001186904.1
NM_001254752.1 4224 UTR 3 NP_001241681.1
NM_006294.4 4224 UTR 3 NP_006285.1

View Full Product Details