Product Details
- SNP ID
-
rs2949865
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:94022066 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCTTGAGTCTACTTAGAATTAGC[G/T]CTTTCACTGGTAAAAAGTTTATTCC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HEPHL1
PubMed Links
Gene Details
- Gene
- HEPHL1
- Gene Name
- hephaestin like 1
View Full Product Details