Product Details
- SNP ID
-
rs12280272
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:129380511 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAGTCAGAGACCCAAACTCATAG[A/G]ATCTTAGGAACCTGAGAGTTGGATA
- Phenotype
-
MIM: 604823
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BARX2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115776058] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BARX2
- Gene Name
- BARX homeobox 2
View Full Product Details