Product Details

SNP ID
rs2288402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:129868456 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTATCAGCACTGCTTAAGGTCTAG[C/G]GGAAACAGGAGGCGTGCTGATGGGG
Phenotype
MIM: 164013
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NFRKB PubMed Links

Gene Details

Gene
NFRKB
Gene Name
nuclear factor related to kappaB binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143835.1 Intron NP_001137307.1
NM_006165.3 Intron NP_006156.2
XM_005271567.3 Intron XP_005271624.1
XM_005271568.3 Intron XP_005271625.1
XM_011542850.2 Intron XP_011541152.1
XM_011542851.2 Intron XP_011541153.1
XM_011542852.2 Intron XP_011541154.1
XM_017017793.1 Intron XP_016873282.1
XM_017017794.1 Intron XP_016873283.1
XM_017017795.1 Intron XP_016873284.1
XM_017017796.1 Intron XP_016873285.1
XM_017017797.1 Intron XP_016873286.1
XM_017017798.1 Intron XP_016873287.1
XM_017017799.1 Intron XP_016873288.1
XM_017017800.1 Intron XP_016873289.1
XM_017017801.1 Intron XP_016873290.1
Gene
TMEM45B
Gene Name
transmembrane protein 45B
There are no transcripts associated with this gene.

View Full Product Details