Product Details

SNP ID
rs35066734
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:186443781 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAATAATGTTGCTGAAGACAGCT[C/T]TGTTTTGTAGTCAAGCATTGTATCA
Phenotype
MIM: 171490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724919 PubMed Links

Gene Details

Gene
LOC102724919
Gene Name
uncharacterized LOC102724919
There are no transcripts associated with this gene.

Gene
PDC
Gene Name
phosducin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002597.4 1013 UTR 3 NP_002588.3
NM_022576.3 1013 UTR 3 NP_072098.1
XM_011509603.2 1013 UTR 3 XP_011507905.1

View Full Product Details